Learn More
Apexbio Technology LLC Nateglinide 105816-04-4 10mg
SDP
Catalog No. 502240806
Encompass_Preferred
Nateglinide (CAS 105816-04-4) is a synthetic oral antihyperglycemic agent classified within the meglitinide class It functions as an insulin secretagogue by selectively binding to pancreatic -cell ATP-dependent potassium channels resulting in membrane depolarization and subsequent stimulation of insulin release Nateglinide is widely employed in research focused on non-insulin-dependent diabetes mellitus providing a tool for investigating glucose-stimulated insulin secretion -cell responsiveness and the pharmacological modulation of insulinotropic pathways in metabolic disease models
This item is not returnable. View return policy
Catalog No. 50-224-0806 Supplier Apexbio Technology LLC Supplier No. B219810
Nateglinide (CAS 105816-04-4) is a synthetic oral antihyperglycemic agent classified within the meglitinide class It functions as an insulin secretagogue by selectively binding to pancreatic -cell ATP-dependent potassium channels resulting in membrane depolarization and subsequent stimulation of insulin release Nateglinide is widely employed in research focused on non-insulin-dependent diabetes mellitus providing a tool for investigating glucose-stimulated insulin secretion -cell responsiveness and the pharmacological modulation of insulinotropic pathways in metabolic disease models