Learn More
Selleck Chemical LLC Nateglinide 25mg 105816-04-4 A-4166
SDP
Supplier:  Selleck Chemical LLC S248925MG
Encompass_Preferred
Nateglinide (A-4166) is an insulin secretagog agent that lowers blood glucose levels by stimulating insulin secretion from the pancreas. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-618-0