Learn More
Medchemexpress LLC Nateglinide-d5 | 1227666-13-8 | 98.0% | 1 MG
SDP
Catalog No. 5000409073
Encompass_Preferred
Nateglinide-d5 is a deuterium labeled Nateglinide. Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. It inhibits ATP-sensitive K+ channels in pancreatic β-cells and is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09073 Supplier Medchemexpress LLC Supplier No. HYB0422S1MG
Nateglinide-d5 is a deuterium labeled Nateglinide. Nateglinide, a D-phenylalanine derivative, is an orally active and short-acting insulinotropic agent and a DPP IV inhibitor. It inhibits ATP-sensitive K+ channels in pancreatic β-cells and is used for the treatment of type 2 (non-insulin-dependent) diabetes mellitus.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS