Learn More
Cayman Chemical Nateglinide, Purity: ≥98%, CAS: 105816-04-4, 1 g
SDP
Catalog No. 502964463
Encompass_Preferred
A hygoglycemic agent; induces insulin and somatostatin release from perfused rat pancreas from 0.03-3 μM; increases intracellular calcium levels in isolated rat pancreatic β cells at 3 μM; reduces blood glucose levels by 20% in fasted mice at 1.6 mg/kg orally; decreases blood glucose levels in an oral glucose tolerance test in normal rats, genetically diabetic KK mice, and an STZ-induced rat models of diabetess
Catalog No. 50-296-4463 Supplier Cayman Chemical Supplier No. 233201 G
A hygoglycemic agent; induces insulin and somatostatin release from perfused rat pancreas from 0.03-3 μM; increases intracellular calcium levels in isolated rat pancreatic β cells at 3 μM; reduces blood glucose levels by 20% in fasted mice at 1.6 mg/kg orally; decreases blood glucose levels in an oral glucose tolerance test in normal rats, genetically diabetic KK mice, and an STZ-induced rat models of diabetess