Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Naththoxyacetic Acid, free acid, 99.1%, MP Biomedicals™

Catalog No. ICN15578925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15578925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15578925 Supplier MP Biomedicals, Inc Supplier No. 0215578925

missing translation for 'validMainframeMsg'

beta-Naphthoxyacetic Acid is a plant growth regulator, which promote the growth of roots on clippings and prevent fruit from falling prematurely. Soluble in alcohol, ehter and acetic acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-23-0
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004066
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
PubChem CID 8422
ChEBI CHEBI:50397
IUPAC Name 2-naphthalen-2-yloxyacetic acid
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 202.2
Physical Form Crystalline Powder
Chemical Name or Material β-Naththoxyacetic Acid, Free Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.