Learn More
Thermo Scientific™ 20mm Crimp Seals with Septa for Headspace Vials
Catalog No. 0339747 Shop All Thermo Scientific Products
Change view
Click to view available options
Septum:
Aluminum Foil/White High Temp. Silicone
Product Type:
Pressure Release
Catalog No. Septum Product Type
03-397-47 Aluminum Foil/White High Temp. Silicone Pressure Release
1 options
Catalog No. 03-397-47 Supplier Thermo Scientific™ Supplier No. C402037AP
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
20mm Crimp seals with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
Ordering Information:
Septa and crimp seals also sold separately.
Note:
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
TRUSTED_SUSTAINABILITY
Specifications
Description Aluminum Foil/White High Temp. Silicone septa
Material Aluminum, Silicone
Septum Aluminum Foil/White High Temp. Silicone
Product Line National
Closure Size 20 mm
For Use With (Equipment) Headspace Vials
Product Type Pressure Release