Learn More
Thermo Scientific™ National Crimp-Top Autosampler Vials with Caps
Catalog No. 03251422 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
03-251-422 Clear
1 options
Catalog No. 03-251-422 Supplier Thermo Scientific™ Supplier No. 60180593
Thermo Scientific™ National Crimp-Top Autosampler Vials with Caps are offered as clear borosilicate glass 33 with silicone/PTFE seal.
Thermo Scientific™ National Crimp-Top Autosampler Vials with Caps are offered as clear borosilicate glass 33 with silicone/PTFE seal.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Description Blue crimp cap with blue silicone /red PTFE seal
Color Clear
Material Borosilicate Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum silicone /red PTFE seal
Volume (Metric) Working 2 mL
Closure Type Crimp Cap
Quantity 1000/Pk.