Learn More
National Diagnostics™ Calci-Clear™ Decalcifying Solution
SDP
Catalog No. 5089990139
Encompass
Change view
Click to view available options
Quantity:
1 qt.
1 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-899-90139 1 qt.
50-899-90138 1 gal.
50-899-90140 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-899-90139 Supplier National Diagnostics™ Supplier No. HS1041QRT
Add to Cart
Add to Cart
A solution of efficient chelating and sequestering agents used with tissue samples to remove and bind all calcium and other metal ions
  • Moderate speed decalcifier
  • Mildness balanced with performance
  • Reduces interference with subsequent staining
  • Minimizes tissue distortion and structure loss
  • May be used to decalcify any tissue
Specifications
Product Type Decalcifying Solution
Quantity 1 qt.