Learn More
National Diagnostics™ Calci-Clear™ Rapid Decalcifying Solution
SDP
Catalog No. 5089990143
Encompass
Change view
Click to view available options
Quantity:
1 qt.
1 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-899-90143 5 gal.
50-899-90142 1 qt.
50-899-90141 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-899-90143 Supplier National Diagnostics™ Supplier No. HS1055GLL
High-speed decalcifier that permits the processing of tissue samples in considerably less time
  • High speed decalcification
  • Minimizes loss of tissue structure
  • Excellent staining characteristics
Specifications
Product Type Decalcifying Solution
Quantity 5 gal.