Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Shop All Thermo Scientific ProductsDescription
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
- Stainless steel construction
- Autoclavable
Specifications
Specifications
| Description | Manual Crimper |
| No. per Pack | 1 Ea. |
| For Use With (Equipment) | Chromatography, Serum Vial |
| Size | 20 mm |
| For Use With (Application) | Industrial Chromatography |
| Product Line | National |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.