Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Barricade Tapes

Catalog No. 179862F Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black, Yellow
Orange, Black
Red
Red, Black
Displayed Text:
Caution
Caution Biohazard
Caution Construction Area
Chemical Spill
Danger
Do Not Enter
Hazardous Area
Radiation Area
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text
17-986-2F Black, Yellow Chemical Spill
17-981-64A Black, Yellow Caution
17-981-64D Red, Black Danger
17-981-64F Red, Black Do Not Enter
17-981-64B Black, Yellow Caution Construction Area
17-985-484E Black, Yellow Hazardous Area
17-981-64G Orange, Black Caution Biohazard
17-981-64H Red Radiation Area
17-981-64C Black, Yellow Do Not Enter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-986-2F Supplier National Marker™ Supplier No. PT42

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quick and effective way to create a visual fortress around temporary hazards

  • Lightweight tape can be tied, stapled or nailed to posts, fences or metal barricades
  • Continuously repeating messages
  • Polyethylene

Order Info

Roll size: 3 x 1000 in. (7.6 x 2540cm).

Specifications

Dimensions (L x W) 1000 ft. x 3 in.
Color Black, Yellow
Displayed Text Chemical Spill
For Use With (Application) Outdoor or indoor application
Length (English) 1000 ft.
Length (Metric) 304.8 m
Width (English) 3 in.
Width (Metric) 7.62 cm
Material HDPE
Type Barricade Tape
Thickness 3 mil
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.