Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard: Authorized Personnel Only Sign

Catalog No. 18999757 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-999-757 Vinyl
18-999-756 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-757 Supplier National Marker™ Supplier No. SA165P

missing translation for 'validMainframeMsg'

Sign for hazardous areas are emphasized by graphic

  • Orange sign with black text and graphic
  • Available in rigid plastic or pressure-sensitive vinyl
  • Size: 17.7 × 43.2cm (7 × 17 in.)

Specifications

Certifications/Compliance OSHA
Material Vinyl
Quantity 1 Ea.
Height (English) 7 in.
Width (English) 17 in.
Displayed Text BIOHAZARD AUTHORIZED PERSONNEL ONLY
Product Type Safety Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.