Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard: Caution Biological Hazard Sign

Catalog No. 18999758 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-999-758 Vinyl
17-984-10A Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-758 Supplier National Marker™ Supplier No. SA52P

missing translation for 'validMainframeMsg'

Emphasizes safety message with the use of a biohazard graphic

  • Orange sign with black text and graphic
  • Available in rigid plastic or pressure-sensitive vinyl
  • Size: 17.7 × 43.2cm (7 in. × 17 in.)

Specifications

Certifications/Compliance OSHA
Material Vinyl
Quantity 1 Ea.
Height (English) 7 in.
Width (English) 17 in.
Displayed Text BIOHAZARD CAUTION BIOLOGICAL HAZARD/Biohazard Pictogram
Product Type Biohazard Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.