Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Biohazard DOT Shipping Labels

Catalog No. 17987172b Shop All Fisherbrand Products
Change view
Click to view available options
Size (Metric):
10.16 x 10.16 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
17-987-172B 10.16 x 10.16 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-172B

Mark hazardous materials for transport on public highways

Comply with DOT signage regulations.

  • Pressure-sensitive vinyl
  • Color: Black text and graphic on white background
  • Size: 10.16 × 10.16cm (4 × 4 in.)

Specifications

Format Label
Material Vinyl
Product Type DOT Placards and Label
For Use With (Application) Biohazard
Quantity 25/Pk.
Size (English) 4 x 4 in.
Size (Metric) 10.16 x 10.16 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.