Learn More
Description
- Orange sign with black text
- Available in rigid plastic or pressure-sensitive vinyl Sized 17.7 × 17.7cm (7 × 7 in.)
Specifications
Specifications
| Material | Vinyl |
| Quantity | 1 Ea. |
| Height (English) | 7 in. |
| Width (English) | 7 in. |
| Displayed Text | BIOHAZARD |
| Product Type | Biohazard Sign |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.