Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Biohazard: Use Universal Precautions Sign

Catalog No. 18999760 Shop All AccuformNMC Products
Change view
Click to view available options
Width (English):
17 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
18-999-760 17 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-760 Supplier National Marker™ Supplier No. S95R

missing translation for 'validMainframeMsg'

Sign for hazardous areas advising standard precautions

  • Orange sign with black text
  • Available in rigid plastic or pressure-sensitive vinyl
  • Sized 17.7 × 17.7 cm (7 × 7 in.)

Specifications

Material Vinyl
Quantity 1 Ea.
Height (English) 7 in.
Width (English) 17 in.
Displayed Text BIOHAZARD Use Universal Precautions
Product Type Biohazard Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.