Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Caution Hazardous Material Storage Area Signs

Catalog No. 179821J Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
10 x 14 in.
Material:
Aluminum
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
17-982-1J 10 x 14 in. Aluminum
17-985-49A 7 x 10 in. Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-982-1J Supplier National Marker™ Supplier No. C310RB

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Alert employees and guests of hazardous areas in a facility

  • Yellow and black caution sign
  • Available in aluminum and pressure-sensitive vinyl
  • Comes in two sizes

1798549A: PS Vinyl; 17.7 x 25.4cm (7 x 10 in.)
179821J: Aluminum; 25.4 x 35.6cm (10 x 14 in.)

Compliance

Comply with OSHA 29 CFR 1910.196

Specifications

Certifications/Compliance OSHA 29 CFR 1910.196
Material Aluminum
Quantity 1 Ea.
For Use With (Application) Indoor and outdoor use
Height (English) 10 in.
Max. Temperature (English) 183°F
Max. Temperature (Metric) 84°C
Thickness 0.05 in.
Width (English) 14 in.
Dimensions (L x W) 10 x 14 in.
Displayed Text CAUTION Hazardous Material Storage Area
Product Type Caution Sign
Color Black on Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.