Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Chemical Storage Area Walk-On Floor Sign

Catalog No. 18999906 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-999-906 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-906 Supplier National Marker™ Supplier No. WFS19

missing translation for 'validMainframeMsg'

Specifications

Material Vinyl
Quantity 1 Ea.
For Use With (Application) Floor applications to point out safety hazards in the work place
Displayed Text Legend Chemical Storage Area
Product Type Floor Sign
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.