Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Confined Space Keep Out Signs

Catalog No. 18999706 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
10 x 14 in.
Material:
Plastic
Vinyl
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
18-999-706 7 x 10 in. Vinyl
18-999-704 7 x 10 in. Plastic
18-999-705 10 x 14 in. Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-999-706 Supplier National Marker™ Supplier No. D372P

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Vinyl
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Danger signs should be used when an immediate hazard exists which pose a high probability of severe injury
Height (English) 7 in.
Max. Temperature (English) 178°F
Max. Temperature (Metric) 81°C
Thickness 0.004 in.
Width (English) 10 in.
Dimensions (L x W) 7 x 10 in.
Displayed Text DANGER Confined Space Keep Out
Product Type Safety Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.