Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Danger: Confined Space Do Not Enter Sign

Catalog No. 19035024 Shop All AccuformNMC Products
Catalog No. 19-035-024 Supplier National Marker™ Supplier No. D360PB

missing translation for 'validMainframeMsg'

Alerts employees and guests to confined areas

  • Red, white and black sign with white and black text
  • Made of pressure-sensitive vinyl
  • Size: 25.4 × 35.6cm (10 × 14 in.)

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Vinyl
Quantity 1 Ea.
For Use With (Application) Perfect way to alert employees and guests when a hazard is present in your facility
Height (English) 10 in.
Max. Temperature (English) 178°F
Max. Temperature (Metric) 81°C
Thickness 0.05 in.
Width (English) 14 in.
Displayed Text DANGER Permit Required Confined Space Do Not Enter
Product Type OSHA Safety Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.