Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Danger: Follow Confined Space Entry Procedure Before Signs

Catalog No. 18999732 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
25.4 x 35.6 cm
7 x 10 in.
Material:
Plastic
Vinyl
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
18-999-732 7 x 10 in. Vinyl
18-999-731 25.4 x 35.6 cm Plastic
18-999-730 7 x 10 in. Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-999-732 Supplier National Marker™ Supplier No. D277P

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Vinyl
Quantity 1 Ea.
For Use With (Application) Perfect way to alert employees and guests when a hazard is present in your facility
Height (English) 7 in.
Max. Temperature (English) 178°F
Max. Temperature (Metric) 81°C
Thickness 0.004 in.
Width (English) 10 in.
Dimensions (L x W) 7 x 10 in.
Displayed Text DANGER Follow Confined Space Entry Procedure Before
Product Type OSHA Safety Sign
Color Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.