Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Hazardous Material Pressure Sensitive Numbers for Labeling

Catalog No. 1798158E Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
1
2
3
4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-981-58E 4
17-981-58A -
17-981-58B 1
17-981-58C 2
17-981-58D 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-981-58E Supplier National Marker™ Supplier No. DCN44

missing translation for 'validMainframeMsg'

Specifications

Color Black, White
Format Label
Material Vinyl
Product Type Label Number
Display Text 4
For Use With (Application) Hazardous materials
Quantity 5/Pk.
Thickness (English) 0.0045 mil
Width (English) 5 in.
Height (English) 5 in.
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.