Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Keep Aisles Clear Walk-On Floor Sign

Catalog No. 18999896 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-999-896 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-896 Supplier National Marker™ Supplier No. WFS12

missing translation for 'validMainframeMsg'

Specifications

Material Plastic
Quantity 1 Ea.
Diameter (English) 17 in.
For Use With (Application) Essential tool for alerting your patrons where there is a wet floor
Height (English) 24 in.
Width (English) 12 in.
Displayed Text Keep Aisles Clear
Product Type Floor Sign
Color Green on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.