Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Paper Shipping Labels for Hazardous and Nonhazardous Waste

Catalog No. 17985409D Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Hazardous Waste
Hazardous Waste (California)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
17-985-409D Hazardous Waste (California)
17-981-62C Hazardous Waste
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-985-409D Supplier National Marker™ Supplier No. HW16

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical- and weather-resistant labels

Order Info

Measure 6 x 6 in.; Pressure-sensitive paper; Roll of 500

Specifications

Certifications/Compliance EPA and DOT, 17-985-409C or -409D, 17-985-409E
Color Yellow
Format Label
Material Paper
Product Type Label
Display Text Hazardous Waste (California)
For Use With (Application) Hazardous waste
Quantity 500/Pk.
Thickness (English) 4 mil
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.