Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Stick-on Numbers and Symbols for Vinyl and Paper Write-On HMIG Labels

Catalog No. 1798161D Shop All AccuformNMC Products
Change view
Click to view available options
Color:
White, Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-981-61D White, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-981-61D Supplier National Marker™ Supplier No. HM41

missing translation for 'validMainframeMsg'

Stick-on personal protection numbers and symbols made from pressure-sensitive vinyl printed with fade-resistant UV ink

  • For use with 17-981-61 Series HMIG labels and with 19-035-033 Write-On Labels.
  • Place appropriate hazard number and symbol directly on the label

Order Info

Size of personal protection numbers: 0.5 x 0.5 in. (1 x 1cm). Size of personal protection symbols: 0.75 x 0.75 in. (2 x 2cm). Available in pack of 10 sheets.

Specifications

Color White, Blue
Format Label
Material Vinyl
Product Type Label
For Use With (Equipment) 17-981-61 Series HMIG labels and with write on labels
Quantity 10/Pk.
Width (English) 4 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.