Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Stop Signs

Catalog No. 17985482 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-985-482 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-482 Supplier National Marker™ Supplier No. TM13H

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance MUTCD
Material Aluminum
Quantity 1 Ea.
For Use With (Application) For communicating parking and traffic messages to vehicles on the road and at your facility
Height (English) 24 in.
Thickness 0.06 in.
Width (English) 24 in.
Shape Octagonal
Displayed Text Stop
Product Type Stop Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.