Learn More
Description
- Resist water and humidity
- Printed with fade-resistant UV ink
Order Info
Vinyl labels, sized 6 x 4 in. (15 x 10cm), available with or without squares for personal protection symbols.
Specifications
Specifications
| Certifications/Compliance | 29 CFR 1910.1200 |
| Color | Red, Yellow, Blue, White |
| Format | Label |
| Material | Vinyl |
| Product Type | Safety Label Without symbol squares |
| Display Text | Health, Flammability, Reactivity, Personal Protection |
| For Use With (Application) | Hazardous materials |
| Quantity | 10/Pk. |
| Width (English) | 4 in. |
| Height (English) | 6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.