Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Vinyl Write-On HMIG Labels

Catalog No. 1798161C Shop All AccuformNMC Products
Change view
Click to view available options
Product Type:
Safety Label With symbol squares
Safety Label Without symbol squares
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-981-61C Safety Label With symbol squares
17-981-61A Safety Label Without symbol squares
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-981-61C Supplier National Marker™ Supplier No. HM36

missing translation for 'validMainframeMsg'

Ideal for indoor use

  • Resist water and humidity
  • Printed with fade-resistant UV ink

Order Info

Vinyl labels, sized 6 x 4 in. (15 x 10cm), available with or without squares for personal protection symbols.

Specifications

Certifications/Compliance 29 CFR 1910.1200
Color Red, Yellow, Blue, White
Format Label
Material Vinyl
Product Type Safety Label With symbol squares
Display Text Health, Flammability, Reactivity, Personal Protection
For Use With (Application) Hazardous materials
Quantity 10/Pk.
Width (English) 4 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.