Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Write-On Hazardous Communication Labels

Catalog No. 19035033 Shop All AccuformNMC Products
Change view
Click to view available options
Width (English):
6 in.
10 in.
15 in.
Quantity:
1 Ea.
10/Pk.
5/Pk.
Height (English):
6 in.
10 in.
15 in.
Height (Metric):
15.24 cm
25.4 cm
38.1 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Quantity Height (English) Height (Metric)
19-035-033 6 in. 10/Pk. 6 in. 15.24 cm
17-984-5A 10 in. 5/Pk. 10 in. 25.4 cm
17-984-5B 10 in. 1 Ea. 10 in. 25.4 cm
17-984-5D 15 in. 1 Ea. 15 in. 38.1 cm
17-984-5C 15 in. 5/Pk. 15 in. 38.1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-035-033 Supplier National Marker™ Supplier No. HM42

missing translation for 'validMainframeMsg'

For customizing labels

  • Pressure-sensitive vinyl labels
  • Write on and/or use the Personal Protection Numbers and Symbols (Cat. No. 17-981-61D)
  • Measure 6 × 6 in. (15.2 × 15.2cm)

Order Info

Available in pack of 10.

Compliance

Comply with 29 CFR1910.1200.

Specifications

Certifications/Compliance 29 CFR 1910.1200
Color Red, Yellow, Blue, White
Format Label
Material Vinyl
Product Type Label
For Use With (Application) Hazardous materials
Quantity 10/Pk.
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.