Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Write-On NFPA Labels

Catalog No. 1798163B Shop All AccuformNMC Products
Change view
Click to view available options
Width (English):
2 in.
4 in.
Height (English):
2 in.
4 in.
Height (Metric):
5.08 cm
10.16 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English) Height (Metric)
17-981-63B 4 in. 4 in. 10.16 cm
17-981-63A 2 in. 2 in. 5.08 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-981-63B Supplier National Marker™ Supplier No. HMS14L

missing translation for 'validMainframeMsg'

Ideal for use in laboratories and other indoor work areas.

  • Identify specific hazards, as well as the degree of health hazard, flammability, and reactivity associated with chemicals
  • Simply determine the hazards of a specific chemical and write the appropriate NFPA hazard number in the corresponding colored area

Order Info

Two sizes: 2 x 2 in. (5 x 5cm) and 4 x 4 in. (10 x 10cm). Available in rolls of 250 pressure-sensitive paper labels per roll.

Specifications

Color Red, Yellow, Blue, White
Format Label
Material Paper
Product Type Label
Display Text NFPA
For Use With (Application) Use in laboratories and other indoor work areas
Quantity 250/Pk.
Width (English) 4 in.
Height (English) 4 in.
Height (Metric) 10.16 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.