Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Write-On NFPA Labels

Catalog No. p-189570 Shop All AccuformNMC Products
Click to view available options
Width (English):
2 in.
4 in.
Height (English):
2 in.
4 in.
Height (Metric):
5.08 cm
10.16 cm

Catalog No. 17-981-63A

Supplier: National Marker™ HMS12L

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

Ideal for use in laboratories and other indoor work areas.

  • Identify specific hazards, as well as the degree of health hazard, flammability, and reactivity associated with chemicals
  • Simply determine the hazards of a specific chemical and write the appropriate NFPA hazard number in the corresponding colored area

Order Info

Two sizes: 2 x 2 in. (5 x 5cm) and 4 x 4 in. (10 x 10cm). Available in rolls of 250 pressure-sensitive paper labels per roll.

Specifications

Color Red, Yellow, Blue, White
Format Label
Material Paper
Product Type Label
Display Text NFPA
For Use With (Application) Use in laboratories and other indoor work areas
Quantity 250/Pk.
Width (English) 2 in.
Height (English) 2 in.
Height (Metric) 5.08 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.