Learn More
MSC National Safety Apparel Hard Cap, 20 cal/sq cm, 1 ea
Catalog No. 5000787919
Encompass
This arc protection hard cap is constructed from polycarbonate, featuring a clear, anti-fog window with dimensions of 10 in. high by 20 in. wide. Designed for welding applications, it offers robust protection against electrical arc hazards and includes a slotted adapter for secure headgear adjustment.
  • Arc protection hard cap type
  • Anti-fog window coating
  • Polycarbonate window material
  • White headgear color
  • Clear window color
  • Window height 10 in.
  • Window width 20 in.
  • Hazard risk category 2
  • Adapter for headgear adjustment
  • Complies with NFPA 70E, ASTM F2178, and ANSI Z89.1 standards
  • Maximum arc flash protection of 20 cal/sq. cm
Catalog No. 50-007-87919 Supplier MSC Supplier No. 31651920
This arc protection hard cap is constructed from polycarbonate, featuring a clear, anti-fog window with dimensions of 10 in. high by 20 in. wide. Designed for welding applications, it offers robust protection against electrical arc hazards and includes a slotted adapter for secure headgear adjustment.
  • Arc protection hard cap type
  • Anti-fog window coating
  • Polycarbonate window material
  • White headgear color
  • Clear window color
  • Window height 10 in.
  • Window width 20 in.
  • Hazard risk category 2
  • Adapter for headgear adjustment
  • Complies with NFPA 70E, ASTM F2178, and ANSI Z89.1 standards
  • Maximum arc flash protection of 20 cal/sq. cm