Learn More
National Scientific™ BioStor™ MicroTube Racks
Catalog No. 118451
Change view
Click to view available options
Positions:
24
Catalog No. Positions
11-845-1 24
1 options
Catalog No. 11-845-1 Supplier National Scientific™ Supplier No. RS24 15MT
Safely handle critical biological samples with these aluminum racks
  • Durable, chemical and corrosion-resistant racks have no welded seams and may be frozen or autoclaved
  • Hole diameters and adjustable shelves accommodate a variety of tubes
  • Ideal for bench procedures; several styles are stackable to utilize counter top, under hood or cold storage space more efficiently
Specifications
Material Aluminum
Positions 24