Learn More
National Scientific™ MicroFlex™ Pipet Tips
Catalog No. 1184161
Change view
Click to view available options
Tip Style:
Flat
Catalog No. Tip Style
11-841-61 Flat
1 options
This item is not returnable. View return policy
Catalog No. 11-841-61 Supplier National Scientific™ Supplier No. MN320F LR
This item is not returnable. View return policy
Ideal for loading electrophoresis gels
These microcapillary tips allow positive-displacement accuracy and reproducibility using Pipetman, Eppendorf, and most other single-channel air-displacement pipettors.
  • Sterilized
  • RNase/DNase-free
  • Flexible
  • 33mm long
  • 5μl microcapillary tube sections with beveled ends
  • Volume range: 0.1 to 200μL
Specifications
Volume (Metric) 0.1 to 200 μL
Sterility Sterile
Tip Style Flat
Product Type Pipet Tips
Product Line MicroFlex