Learn More
Thermo Scientific™ National Scientific SepCap™ Polyspring™ Limited Volume Inserts for Waters 96-Position Autosampler
Shop All Thermo Scientific ProductsDescription
Prevent the syringe from bottoming out and damaging the needle with Thermo Scientific™ National SepCap Polyspring Limited-Volume Inserts for Waters 96-Position Autosampler.
- Type I, borosilicate glass
Specifications
Specifications
| Volume (Metric) | 250 μL |
| Style | Polyspring Conical, Silanized |
| Product Line | National SepCap Polyspring |