Learn More
Thermo Scientific™ National Target All-Plastic Disposable Luer-Lok Syringes
Catalog No. 0337729W25 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
03-377-29W25 10 mL
1 options
Catalog No. 03-377-29W25 Supplier Thermo Scientific™ Supplier No. 0337729W25
Use these all-plastic disposable syringes to dispense small volumes of liquids, or to pressure filter through our syringe filters.
  • Choose Luer-Slip or Luer Lock syringes 10 mL capacity
  • Nonsterile
  • Bulk packed
TRUSTED_SUSTAINABILITY
Specifications
Material Polyethylene (Plunger), Polypropylene (Barrel)
Product Type Luer-lok Syringe
Disposable Single-use
Quantity Pack of 100 Syringes
Volume (Metric) 10 mL
Product Line National Target
For Use With (Equipment) Syringe Filter
Connection Type Luer Lock
Sterility Non-sterile