Learn More
Thermo Scientific™ National Target All-Plastic Luer-Lok Syringes
Catalog No. 0337728W25 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
5 mL
Catalog No. Volume (Metric)
03-377-28W25 5 mL
1 options
Catalog No. 03-377-28W25 Supplier Thermo Scientific™ Supplier No. 0337728W25
Use these all-plastic disposable syringes to dispense small volumes of liquids, or to pressure filter through our syringe filters.
  • Choose Luer-Slip or Luer Lock syringes 5 mL capacity
  • Nonsterile
  • Bulk packed
TRUSTED_SUSTAINABILITY
Specifications
Material Polyethylene (Plunger), Polypropylene (Barrel)
Product Type Luer-lok Syringe
Disposable Single-use
Volume (Metric) 5 mL
Product Line National Target
For Use With (Equipment) Syringe Filter
Connection Type Luer Lock
Sterility Non-sterile