Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Catalog No. 03376430 Shop All Thermo Scientific Products
Change view
Click to view available options
Marking Spot:
No
Yes
Closure Color:
Black
Light Blue
Septum:
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Solid PTFE
Color:
Amber
Clear
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Marking Spot Closure Color Septum Color
03-376-430 No Light Blue PTFE/Silicone, Pre-slit Clear
03-375-30A No Light Blue PTFE/Silicone Clear
03-376-431 No Black PTFE/Silicone Amber
03-376-433 Yes Light Blue PTFE/Silicone/PTFE Amber
03-376-434 Yes Light Blue Solid PTFE Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-376-430 Supplier Thermo Scientific™ Supplier No. C4010018
Get 1 Extra Pack of Vials When You Buy 2
View all promotions
Add to Cart
Add to Cart
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Light Blue
Description Clear Glass; Light Blue Cap; PTFE/Silicone Pre-slit septum
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Pre-slit Yes
Septum PTFE/Silicone, Pre-slit
Marking Spot No