Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National Target™ Empty Caps without Septum for 10-425 Vials

Catalog No. 03376473 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Green
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-376-473 Yellow
03-376-446 Green
03-376-447 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-376-473 Supplier Thermo Scientific™ Supplier No. C401098Y

missing translation for 'validMainframeMsg'

Thermo Scientific™ National Target Empty Open-Top Caps for 10-425 Vials are designed to be used with any of our 10mm septa.

Thermo Scientific™ National Target Empty Open-Top Caps for 10-425 Vials are designed to be used with any of our 10mm septa.

  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Target 10-425 caps feature a flanged rim, making them suitable for Japanese autosampler trays
TRUSTED_SUSTAINABILITY

Specifications

Description Yellow; For 10-425 Target Vials
Color Yellow
Material Polypropylene
Product Line National
Closure Size 10-425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.