Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National™ Target™ Kim-Snap™ Closures

Catalog No. 03396E Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Yellow
Material (Liner):
PTFE/Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material (Liner)
03-396E Yellow PTFE/Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-396E Supplier Thermo Scientific™ Supplier No. C401151Y

missing translation for 'validMainframeMsg'

Preassembled with septa for use with Target Snap/Crimp LoVial™ Vials

  • Effective evaporation resistance—apply with simple thumb pressure
  • Replace aluminum crimp seals and end tedious crimping/de-crimping

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) PTFE/Rubber
Color Yellow
Material Polypropylene
Closure Size 11 mm
For Use With (Equipment) Target Snap/Crimp LoVial Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.