Description
Thermo Scientific™ National Target Snap-It Storerooms provide convenient storage for vials and caps.
- Mini Storeroom has 6 drawers, each drawer will hold one 100/pack of vials or 500/pack caps
- Full-size Storeroom has 9 deep drawers with a capacity of 2000 vials and caps
- Storeroom can be restocked as product is consumed
Specifications
Specifications
| Closure Color | Clear |
| Description | NSC Storeroom, Clear Vial, PTFE/RR, Clear Snap-It Cap, 2000 pieces |
| Material | PTFE |
| Septum | PTFE/RR |
| Closure Material | PTFE, Rubber |
| Closure Type | Snap-It Cap |