Learn More
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Catalog No. p-4526340
Change view
Click to view available options
Marking Spot:
No
Yes
Septum:
PTFE
PTFE/Silicone, Pre-slit
Color:
Clear
Material:
Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Marking Spot Septum Color Material
03-376-443 Yes PTFE Clear Glass
03-378-04 No PTFE/Silicone, Pre-slit Clear Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-376-443 Supplier Thermo Scientific™ Supplier No. C401090W
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
  • Two-compartment benchtop storage tray
  • I-D™ vials feature write-on patch with graduations
  • All items remain handy and dust-free
  • Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Light Blue
Description Clear Glass; ID patch; Light Blue Cap, White PTFE Septum; 2mL
Color Clear
Material Glass
Pre-slit No
Volume (Metric) 2 mL
Septum PTFE
Marking Spot Yes