Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Catalog No. 03378370 Shop All Thermo Scientific Products
Change view
Click to view available options
Marking Spot:
No
Yes
Septum:
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Color:
Amber
Clear
Color (Seal):
Red
Red/Clear
Red/White
Red/White/Red
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Marking Spot Septum Color Color (Seal)
03-378-370 Yes PTFE/Silicone Clear Red/White
22-045922 No PTFE/Silicone/PTFE Clear Red/White/Red
22-045926 No PTFE/Silicone Clear Red/Clear
03-378-371 No PTFE/Silicone Amber Red/White
03-378-368 No PTFE/Rubber Clear Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-378-370 Supplier Thermo Scientific™ Supplier No. C4013492W
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
  • Two-compartment, see-through benchtop storage tray
  • Caps with septa are not attached to vials (packaged in plastic bags)
Includes: 
100 vials and 100 caps with pre-assembled septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color (Seal) Red/White
Description Vials: Clear I-D; Closure: Black Phenolic; Septa: Red PTFE/White Silicone
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Septum PTFE/Silicone
Closure Type Phenolic
Marking Spot Yes