Learn More
Thermo Scientific™ WebSeal™ Microplate Sealing System
Catalog No. 0339784 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Wells:
96
Well Shape:
Round
Square
Color:
Pink
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Well Shape Color
03-397-84 96 Square Yellow
03-397-81 96 Round Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-397-84 Supplier Thermo Scientific™ Supplier No. M9601SQ2Y
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
When productivity really counts and cross-contamination simply won't be tolerated, choose the WebSeal System for efficiency and security. WebSeal system products are used in high-throughput screening, combinatorial chemistry, life science and HPLC. This system is a comprehensive range of 96-well microtiter plates with glass inserts and silicone/PTFE sealing mats.
  • Manufactured from pure silicone with PTFE coating on underside
  • Excellent chemical compatibility
  • Dry heat autoclavable
  • Eliminates cross-contamination well to well
  • Available pre-slit for easy penetration and reduced vacuum formation
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material PTFE-coated Silicone
No. of Wells 96
No. per Pack 10
Product Type Microplate Sealing Components
Product Line Webseal
Well Shape Square
Plate Compatibility 96-well Plate