Learn More
Creative Biomart Native Angiostatin
Catalog No. 502084298
Encompass_Preferred
Angiostatin is an internal proteolytic fragment of plasminogen, containing at least three Kringle domains.
Catalog No. 50-208-4298 Supplier Creative Biomart Supplier No. THP0247
Angiostatin is an internal proteolytic fragment of plasminogen, containing at least three Kringle domains.