Learn More
Crescent Chemical Co Inc NATIVE ANODE BUFFER BLUE/CLEAR
SDP
Catalog No. 50248598
Encompass_Preferred
Native Anode Buffer for Blue/ Clear Native (10x) Running buffer for the use as anode buffer in Blue or Clear Native PAGE. Supplied as 10x concentrate. Contains 500 mM BisTris-HCl (pH 7.0). Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42535.01
Catalog No. 50-248-598 Supplier Crescent Chemical Co Inc Supplier No. SE42535.01
Native Anode Buffer for Blue/ Clear Native (10x) Running buffer for the use as anode buffer in Blue or Clear Native PAGE. Supplied as 10x concentrate. Contains 500 mM BisTris-HCl (pH 7.0). Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42535.01