Learn More
Crescent Chemical Co Inc NATIVE CATHODE BUFFER FOR BLUE
SDP
Catalog No. 50248599
Encompass_Preferred
Native Cathode Buffer for Blue/ Clear Native (10x) Running buffer for the use as cathode buffer in Blue or Clear Native PAGE. Supplied as 10x concentrate. Contains 500 mM Tricine, 150 mM BisTris. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 42536.01 CAS N
Catalog No. 50-248-599 Supplier Crescent Chemical Co Inc Supplier No. SE42536.01
Native Cathode Buffer for Blue/ Clear Native (10x) Running buffer for the use as cathode buffer in Blue or Clear Native PAGE. Supplied as 10x concentrate. Contains 500 mM Tricine, 150 mM BisTris. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 42536.01 CAS N