Learn More
Abcam Native Clostridium difficile Toxoid B protein, 50UG
Catalog No. 502584506
Encompass_Preferred
Native Clostridium difficile Toxoid B protein is a Clostridium difficile Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, ELISA.
Protein/ Peptide/ Enzyme Synonyms: toxB, tcdB, Toxin B
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4506 Supplier Abcam Supplier No. AB12400250UG
Native Clostridium difficile Toxoid B protein is a Clostridium difficile Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, ELISA.
Protein/ Peptide/ Enzyme Synonyms: toxB, tcdB, Toxin B
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement