Learn More
Abcam Native Dog Canine Albumin protein, 100MG
Catalog No. NC1829277
Encompass_Preferred
Native Dog Canine Albumin protein is a Dog Full Length protein, in the 25 to 608 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Albumin, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1829277 Supplier Abcam Supplier No. AB119814100MG
Native Dog Canine Albumin protein is a Dog Full Length protein, in the 25 to 608 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Albumin, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement