Learn More
Abcam Native Human Apolipoprotein B, 500UG
Catalog No. 502709051
Encompass_Preferred
Native Human Apolipoprotein B is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Apolipoprotein B-100, Apo B-100, APOB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9051 Supplier Abcam Supplier No. AB91112500UG
Native Human Apolipoprotein B is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Apolipoprotein B-100, Apo B-100, APOB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement